Skip to Content
MilliporeSigma

D3447

Doxycycline hydrochloride

Synonym(s):

2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride (1:1), (4S,4aR,5S,5aR,6R,12aS)-

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
MDL number:
UNSPSC Code:
51284028
NACRES:
NA.85
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

synthetic

form

powder

color

yellow

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycoplasma, parasites

mode of action

protein synthesis | interferes

storage temp.

−20°C

InChI

1S/C22H24N2O8.ClH/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-7,10,14-15,17,25-27,31-32H,23H2,1-3H3;1H/b21-13-;/t7-,10+,14+,15-,17-,22-;/m0./s1

InChI key

VLUQVUWDECWBTL-UQVCFKGQSA-N

General description

Doxycycline is a member of the tetracycline antibiotics group, and is commonly used to treat a variety of infections. It is a Broad spectrum antibiotic and bacteriostatic. Doxycycline shows antiprotozoal properties and is a potent inhibitor of MMPs (matrix metalloproteinases) in vivo.

Application

Doxycycline hydrochloride has been used for the induction of shRNA expression and induction of expression vectors.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library