Skip to Content

D1657

1,3-Dioleoyl-2-palmitoylglycerol

≥99%

Synonym(s):

1,3-Di(cis-9-octadecenoyl)-2-hexadecanoylglycerol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C55H102O6
CAS Number:
Molecular Weight:
859.39
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

form

liquid

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC(COC(=O)CCCCCCC\C=C\CCCCCCCC)COC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C55H102O6/c1-4-7-10-13-16-19-22-25-27-30-32-35-38-41-44-47-53(56)59-50-52(61-55(58)49-46-43-40-37-34-29-24-21-18-15-12-9-6-3)51-60-54(57)48-45-42-39-36-33-31-28-26-23-20-17-14-11-8-5-2/h25-28,52H,4-24,29-51H2,1-3H3/b27-25+,28-26+

InChI key

PPTGNVIVNZLPPS-NBHCHVEOSA-N

General description

1,3-Dioleoyl-2-palmitoylglycerol (OPO) is a structured triglyceride in infant formula. Its structure is like human milk fat (HMF).

Application

1,3-Dioleoyl-2-palmitoylglycerol (OPO) has been used:
  • as a synthetic triacylglycerol (TAG) standard to investigate the fragmentation patterns of TAGs in milk fat by the multi-dimension mass spectrometry (MDMS) method
  • as a lipid standard to analyze OPO content/TAG compositions in the acidolysis products by high-performance liquid chromatography-evaporative light scattering detector (HPLC-ELSD)
  • as a standard in silver-ion HPLC separation of OPO-rich human milk fat substitute (HMFS)

Biochem/physiol Actions

1,3-Dioleoyl-2-palmitoylglycerol (OPO) exerts several beneficial effects on infants, including remitting constipation, improved calcium and fatty acid absorption, and improved bone development.

Packaging

Sealed ampule


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library