Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D1639

Sigma-Aldrich

Glyceryl 1,3-dipalmitate

≥99%

Synonym(s):

α,γ-Dipalmitin, 1,3-Dihexadecanoylglycerol, 1,3-Dipalmitin, 1,3-Dipalmitoyl-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H68O5
CAS Number:
Molecular Weight:
568.91
Beilstein:
1809772
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99%

form

powder

impurities

≤1% 1,2-isomer

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCCCCCC

InChI

1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-31-33(36)32-40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3

InChI key

GFAZGHREJPXDMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glyceryl 1,3-dipalmitate has been used as a standard in lipid analysis of feeds and omasal digesta. It has also been used as a standard in identifying sn-1,3 diacylglycerol (DAG) in skeletal muscles.

Biochem/physiol Actions

Glyceryl 1,3-dipalmitate from Lactobacillus paracasei subsp. possesses neuroprotective effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Halmemies-Beauchet-Filleau et al.
Journal of dairy science, 96(9), 5882-5900 (2013-07-16)
Diets based on red clover silage (RCS) typically increase the concentration of polyunsaturated fatty acids (PUFA) in ruminant milk and meat compared with grass silages (GS), an effect that has been attributed to higher activity of polyphenol oxidase in red
Mari Merce Cascant et al.
Analytical and bioanalytical chemistry, 409(14), 3527-3539 (2017-04-09)
There is a great interest in finding alternatives and green solvents in extraction processes to replace petroleum based solvents. In order to investigate these possibilities, computational methods, as Hansen solubility parameters (HSP) and conductor-like screening model for real solvent (COSMO-RS)
Annette Karen Serup et al.
Diabetes, 65(10), 2932-2942 (2016-08-05)
Type 2 diabetes and skeletal muscle insulin resistance have been linked to accumulation of the intramyocellular lipid-intermediate diacylglycerol (DAG). However, recent animal and human studies have questioned such an association. Given that DAG appears in different stereoisomers and has different
Meng-Chun Cheng et al.
Journal of agricultural and food chemistry, 65(36), 7926-7933 (2017-08-23)
Glyceryl 1,3-dipalmitate (GD) purified from Lactobacillus paracasei subsp. paracasei NTU 101-fermented products has been demonstrated to possess neuroprotective properties. We determined the effect of GD on oxygen-glucose deprivation and reperfusion (OGD/R)-induced SH-SY5Y neuroblastoma cell death. GD ameliorated OGD/R-induced apoptosis by
Xiuwen Wang et al.
Biotechnology letters, 41(6-7), 789-799 (2019-05-10)
To investigate the lipase-catalyzed synthesis of high purity sn-1/3 and sn-2 monoacylglycerols (1/3-MAG and 2-MAG) of different fatty acids (FAs). The 1/3-MAGs of three FAs (16:0, 17:0, 16:1) were synthesized using lipase-catalyzed esterification of glycerol with FAs. The 2-MAGs were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service