Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS9102

Corning® 96 well plates

Clear Flat Bottom Polystyrene, TC-Treated, Individually Wrapped, Sterile, 1 x 8 Stripwell

Synonym(s):

cell culture plate, stripwell, tissue culture plates, well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15

material

clear bottom
clear wells
flat wells
polystyrene
polystyrene plate
round wells

description

growth area .32 cm2

sterility

sterile

feature

lid
skirt
1 x 8 Stripwell
plate format: 96 well standard

packaging

case of 100
pack of 25
case of 50 (individually wrapped)

manufacturer/tradename

Corning 9102

size

96 wells

well diam.

6.4 mm

well volume

360 μL
360 μL

well working volume

75-200 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Corning® 96-well (1 x 8 Stripwell) is suitable for cell based assays.

Features and Benefits

  • 8 well strip plate, assembled 12 strips per plate
  • Egg-crate style strip holder allows each individual well to be positioned back into the plate once strip is broken
  • 1 x 8 strips are designed to fit only one way into the strip holder, to avoid the chance of misorientation
  • Flat bottom microplate with total well volume of 360 μL and recommended working volumes of 75 to 200 μL
  • Surface treated for optimal cell attachment and to improve the binding of difficult to attach cell lines
  • Individually wrapped with nonreversible lids containing condensation rings to reduce contamination
  • Microplates are sterilized and certified nonpyrogenic
  • Individual alphanumeric codes enable easy well identification

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tejashree Redij et al.
Journal of computer-aided molecular design, 33(11), 973-981 (2019-11-24)
The Glucagon-like peptide 1 receptor (GLP-1R) is a well-established target for the treatment of type 2 diabetes and GLP-1R agonist-based therapies represent an effective approach which results in several GLP-1 analog drugs. However, the development of nonpeptidic agonist drugs targeting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service