Key Documents
CLS70075100
Pyrex® VISTA graduated cylinder, to contain, economy
volume 100 mL
Synonym(s):
lab equipment accessory
About This Item
UNSPSC Code:
41121805
NACRES:
NB.43
manufacturer/tradename
Corning 70075-100
volume
100 mL
General description
These Pyrex VISTA cylinders are calibrated "to contain" (TC). They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Tolerances are +/- 5% of total at any point. By removing the base, the graduate can be conveniently stacked. Bumper guards are supplied with 25 mL through 100 mL inclusive. The 10 mL has a funnel top for careful pouring.
Legal Information
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)