Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS431430

Corning® square polycarbonate storage bottle

square clear polycarbonate, sterile, bottle capacity 150 mL, cap (plug seal), case of 24

Synonym(s):

polycarbonate containers, polycarbonate plastic containers, polycarbonate storage containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.75

material

cap (plug seal)
polycarbonate bottle (square)
screw top polypropylene cap
square clear polycarbonate

sterility

sterile
sterile

feature

cap
disposable
graduations 50 mL

packaging

case of 24
case of 24

manufacturer/tradename

Corning 431430

availability

not available in Canada

bottle capacity

150 mL

bottle neck diam.

~44 mm

fitting

GL 45 thread

Looking for similar products? Visit Product Comparison Guide

General description

Corning square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.

Caution: These square polycarbonate storage bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

  • Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.
  • Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
  • Screened white enamel graduations are easier to see than molded graduations
  • Sterilized by gamma radiation
  • Large white marking spot for easier identification
  • Bottles can be autoclaved once at 121ºC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.
  • Optional reusable caps with silicone septa (SIAL Cat. Nos. CLS139545HTSC, CLS139545TS) for syringe sampling or introduction of reagents are available for these caps.

Note:
Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service