MilliporeSigma
CLS4080
Corning® Lambda® plus 8-channel pipettor
volume 0.5-10 μL
Synonym(s):
multi-channel pipettor
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
10 μL
Feature:
8-channel
Manufacturer/tradename:
Corning 4080
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
8-channel
manufacturer/tradename
Corning 4080
volume measuring range
0.5-10 μL
volume
10 μL
General description
  • Corning® Lambda® plus 8-channel pipettor is designed for universal fit with all common brands of pipet tips.
  • Factory calibrated to strict ISO 8655 standards-3-year warranty.
  • Smooth plunger movement and extremely low pipetting forces.
  • Fully autoclavable and UV resistant.
  • Adjustable tip ejector.
  • Engineered to provide the highest levels of accuracy, reproducibility, and comfort.
  • Inclusive of key for easy in-house calibration.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)