Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3841

Corning® 96 Well Poly-D-Lysine Treated Microplate

flat bottom clear, clear polystyrene, bag of 20, sterile, lid

Synonym(s):

Poly-D-Lysine 96-Well Plate, Treated 96-Well Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

clear polystyrene
clear wells
flat bottom clear
round wells

sterility

sterile

feature

lid
skirt

packaging

case of 100
bag of 20

manufacturer/tradename

Corning 3841

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

clear

binding type

poly-D-lysine coated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • Coated with poly-D-lysine for enhanced cell attachment
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterile (aseptic assembly) and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Bulk packed 20 per bag

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service