CLS3839BC
Corning® 1536 well plate, with generic barcode
size 1536 wells, black/clear bottom polystyrene wells, low base, PDL, lid, sterile
Synonym(s):
1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, collagen coated plate
About This Item
polystyrene plate
Recommended Products
material
black/clear bottom polystyrene wells
polystyrene plate
sterility
sterile
feature
lid
PDL
low base
packaging
pack of 10 ea
pack of 10
case of 50 ea
case of 50
manufacturer/tradename
Corning 3839BC
maximum volume
12.5 μL
size
1536 wells
working volume
8 μL
suitability
suitable for (cell cuture)
General description
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
- Resistant to temperature ranges of -80ºC to +52ºC
- 2.8 mm distance between well bottoms to plate bottom
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
Microplate Features
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness -minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²)
- Working volume of up to 10 μL
Bar Code Details
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)