Skip to Content
MilliporeSigma

CLS3824BC

Corning® 384 well microplate, low volume

white polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10

Synonym(s):

384 multiwell plate, 384 well barcode plate, 384 well microplate, 384 well microtiter plate, 384 well plate, 384 well plate with generic barcode

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41121800
Material:
flat bottom , white bottom, white plate, white polystyrene
Size:
384 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom , white bottom, white plate, white polystyrene

sterility

non-sterile

feature

lid: no, skirt, with generic barcode

packaging

pack of 10

manufacturer/tradename

Corning 3824BC

size

384 wells

well volume

50 μL

wells

384 wells , low volume

working volume

5-40 μL

color

white

suitability

suitable for (cell cuture)

binding type

NBS (non-binding surface)

General description

Microplate Features
  • Round well design with a solid white flat bottom.
  • 2.8 mm distance between well bottoms to plate bottom.
  • Ideal for miniaturized 384 well fluorescent homogeneous assays.
  • Nonbinding surface (NBS) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions.
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library