Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3590

Corning® 96 Well EIA/RIA Assay Microplate

flat bottom clear, polystyrene, high binding surface, pkg of (individually wrapped), non-sterile, lid: no

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear
polystyrene
polystyrene
round wells
size:
96 wells , 96 well plate

material

flat bottom clear
polystyrene
polystyrene
round wells

sterility

non-sterile

feature

lid: no
skirt
plate format: 96 well standard
well: flat bottom, clear

packaging

pack of 1
case of 100
pkg of (individually wrapped)

manufacturer/tradename

Corning 3590

size

96 wells , 96 well plate

well volume

330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL

well working volume

75-200 μL

color

clear plate

suitability

suitable for (EIA/RIA; immunoassays)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • High binding surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity of approximately 500 ng of mouse IgG/cm2
  • Ideal for immunoassays
  • EIA/RIA plates are tested on lot-to-lot basis and the certification results for each lot are available upon request
  • Recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
  • Without lids, lids supplied separately
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.

Legal Information

Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J Olson et al.
Cell reports, 28(11), 2878-2891 (2019-09-12)
CD4 T follicular helper (Tfh) cells are specialized in helping B cells during the germinal center (GC) reaction and ultimately promote long-term humoral immunity. Here we report that loss of the nuclear orphan receptor NR2F6 causes enhanced survival and accumulation of
Xiangyang Zhang et al.
Journal of immunology (Baltimore, Md. : 1950), 205(7), 1743-1751 (2020-08-26)
Myasthenia gravis (MG) is an autoimmune disease in which Abs target neuromuscular junction proteins, in particular the acetylcholine receptor. We previously identified the antiapoptotic protein survivin in the autoreactive B cells and plasma cells of MG patients. To further define
Katia Schoeler et al.
The FEBS journal, 286(18), 3566-3581 (2019-05-24)
Upon activation by antigen, B cells form germinal centres where they clonally expand and introduce affinity-enhancing mutations into their B-cell receptor genes. Somatic mutagenesis and class switch recombination (CSR) in germinal centre B cells are initiated by the activation-induced cytidine
Karen Cortés-Sarabia et al.
Applied microbiology and biotechnology, 104(14), 6173-6183 (2020-05-29)
Bacterial vaginosis is one of the most frequent vaginal infections. Its main etiological agent is Gardnerella vaginalis, which produces several virulence factors involved in vaginal infection and colonization, in particular, sialidase (SLD), a potential clinical biomarker that participates in immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service