Skip to Content
MilliporeSigma

CLS3574

Corning® NBS 384 well microplates

surface treatment, Non-binding Surface, white polystyrene, flat bottom, non-sterile, lid: no, pack of 10

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41121800
Material:
flat bottom , flat bottom wells white , polystyrene , square white wells, white polystyrene
Size:
384 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom , flat bottom wells white , polystyrene , square white wells, white polystyrene

sterility

non-sterile

feature

lid: no, skirt

packaging

pack of 10, case of 50

manufacturer/tradename

Corning 3574

maximum volume

112 μL

size

384 wells

working volume

80 μL

color

white

suitability

suitable for (cell cuture)

binding type

NBS (non-binding surface)

General description

  • White wall plates are designed to reduce well-to-well crosstalk during luminescent assays
  • Recommended working volume of up to 20 to 80μL
  • Enhanced flatness provides low intra- and inter- plate CV′s.
  • Nonbinding (NBS) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
  • Conforms to standard microplate footprint and dimensions
  • Ideal for today′s more sensitive assays and plate readers
Non-Binding surface (NBS) multiwell plates have a modified polymer surface (PS) which results in a non-ionic hydrophilic surface (polyethylene oxide-like), thereby minimizing bio-interaction and enhancing the signal-to-noise ratio. Results have demonstrated NBS′s ability to significantly reduce protein and nucleic acid binding to polymers (<2 ng/cm2; over 90% reduction of protein binding compare to PS & PP surface), maintain enzyme activity, and inhibit adhesion of a number of cell lines. These novel surface technologies for assay plates are ideal for homogeneous assays such as Scintillation Proximity Assay (SPA). In addition to a 20+ fold increase in signal-to-noise ratio, selected assays have also displayed improved sensitivity and specificity. The recommended working volume of up to 20 μL to 80 μL for 384 well plates.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library