Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS356505

Corning® BioCoat Collagen I 48 Well Clear Flat Bottom TC-Treated Multiwell Plate with Lid

well size 48 wells, flat bottom clear wells, lid

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13
material:
flat bottom clear wells
well size:
48 wells
binding type:
Tissue Culture (TC)-treated surface
feature:
lid

material

flat bottom clear wells

feature

lid

packaging

pkg of 50 ea (10 packs of 50)

manufacturer/tradename

Corning 356505

well size

48 wells

binding type

Tissue Culture (TC)-treated surface

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service