Skip to Content
MilliporeSigma

CLS354522

Corning® BioCoat™ Laminin Cell Culture Flask

capacity 75 cm2, rectangular flask, canted neck, cap, with Plug Seal Cap

Synonym(s):

cell culture flask, plastic cell culture flask, coated flask

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41122104
eCl@ss:
32040201
Binding type:
Tissue Culture (TC)-treated surface
Material:
rectangular flask
Packaging:
pkg of 10 ea (2 packs of 10)
Feature:
cap with Plug Seal Cap, canted neck
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

rectangular flask

feature

cap with Plug Seal Cap, canted neck

packaging

pkg of 10 ea (2 packs of 10)

manufacturer/tradename

Corning 354522

capacity

75 cm2

binding type

Tissue Culture (TC)-treated surface

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library