Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS354232

Corning® Laminin

from mouse, pack of 1 mg

Synonym(s):

Corning Laminin, Laminin Coating

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NA.7

biological source

mouse

packaging

pack of 1 mg

manufacturer/tradename

Corning 354232

General description

Corning Laminin, mouse, 1mg, is used as a thin coating on tissue-culture surfaces or as a soluble additive to culture medium. Laminin is a major component of basement membranes. It has numerous biological activities including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.
Quality
Purity >90% by SDS-PAGE (contains entactin)
Tested for ability to initiate neurite outgrowth of NG-108 human neuroblastoma cells
Found negative for bacteria, fungi, and mycoplasma

Source
Engelbreth-Holm-Swarm (EHS) mouse tumor

Formulation
Frozen in 0.05 M Tris-HCl, 0.15 M NaCl, pH 7.4

Molecular Weight
900 kD (unreduced)

Preparation and Storage
Stable for at least three months at -70°C.

Handling
Recommended concentration: 1-10 μg/cm² of growth surface depending on cell type

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service