Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS352096

Corning® Falcon® Centrifuge Tubes

capacity 15 mL, conical polypropylene, cap, dome seal screw, bagged, sterile

Synonym(s):

Corning Centrifuge Tubes, Falcon Centrifuge Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.13

material

conical polypropylene

sterility

sterile

feature

cap dome seal screw

manufacturer/tradename

Corning 352096

capacity

15 mL

Looking for similar products? Visit Product Comparison Guide

Application

Liquid Handling

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service