Skip to Content
MilliporeSigma

CLS3344

Corning® 96 Well Storage Microplate

V-bottom, expanded volume, polypropylene, standard height, bag of 10 ×, sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121800
Material:
V-bottom , polypropylene
Size:
96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

V-bottom , polypropylene

sterility

sterile

feature

lid: no

manufacturer/tradename

Corning 3344

packaging

bag of 10 ×

maximum volume

450 μL

size

96 wells

working volume

450 μL

color

clear

suitability

suitable for (general storage, liquid handling; automated applications)

binding type

non-treated surface

Features and Benefits

  • Corning® 96 well microplates support various assay and storage application requirements.
  • Features uniform skirt heights for the greater robotic gripping surface.
  • Resistant to most solvents including DMSO, ethanol, methanol.
  • RNase-/DNase-free.

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library