Skip to Content
MilliporeSigma

CLS3336

Corning® CellBIND® Multiple Well Plate

size 12 wells, flat clear bottom, sterile, lid

Synonym(s):

12 well culture plate, cell culture multiwell plates, cell culture plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122107
NACRES:
NB.13
eCl@ss:
32190102
Material:
clear polystyrene plate, flat bottom wells clear , flat clear bottom, polystyrene
Size:
12 wells
Sterility:
sterile
Binding type:
CellBIND® surface
Feature:
lid, skirt, plate format: 12 well standard
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear polystyrene plate, flat bottom wells clear , flat clear bottom, polystyrene

sterility

sterile

feature

lid, skirt, plate format: 12 well standard

packaging

pack of 5 ea, case of 50 ea

manufacturer/tradename

Corning 3336

technique(s)

cell culture: suitable

size

12 wells

surface area

3.8 cm2 , cell growth area

well volume

6.9 mL

well working volume

0.76-1.14 mL

color

clear

suitability

suitable for (cell culture involving anchorage-dependant cell lines)

binding type

CellBIND® surface

General description

Corning® CellBIND® Cell Culture Multiwell Plates

  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library