Skip to Content
MilliporeSigma

CLS3085

Corning® multiwell plates, plate lids and sealing mats

DMSO-resistant cyclic-olefin copolymer universal lid, universal fit, non-sterile, 50/cs

Synonym(s):

microplate lid, multiwell plate lid, universal microplate lid, universal multiwell plate lid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41121800
Material:
amber wells, cyclic olefin copolymer (COC)
Size:
96 or 384 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

amber wells, cyclic olefin copolymer (COC)

sterility

non-sterile

feature

lid

packaging

pack of 25, case of 50

manufacturer/tradename

Corning 3085

size

96 or 384 wells

color

amber

suitability

suitable for (cell cuture)

General description

  • Tinted amber for light-sensitive assays
  • These standard height lids are made of cyclic-olefin which is 100% DMSO-resistant
  • Lids do not fit Corning 2 μL 1536 well microplates
  • Nonsterile

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library