Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS2018

Corning® cap inserts for cryogenic vials

green

Synonym(s):

Corning cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121704
NACRES:
NB.13

sterility

non-sterile

manufacturer/tradename

Corning 2018

color

green

Looking for similar products? Visit Product Comparison Guide

General description

  • Cap inserts provide color coding for easy sample identification
  • Inserts are packaged in resealable bags
  • Cap inserts fit all Corning cryogenic vials except Corning No. 430289

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service