Key Documents
CLS20012
Corning® HYPERStack® 12-layer cell culture vessel
CellBIND® surface, sterile, bag of 1 ea, case of 4 ea
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
material
PTFE filters (membrane)
polycarbonate filling tube
polycarbonate plug (membrane)
polystyrene body (virgin)
polystyrene film
rectangular flask
sterility
sterile
packaging
bag of 1 ea
case of 4 ea
manufacturer/tradename
Corning 20012
technique(s)
cell culture | mammalian: suitable
surface area
6000 cm2 , cell growth area
binding type
CellBIND® surface
General description
The utilization of gas-permeable film technology allows Corning® High Yield PERformance (HYPER) cell culture vessels to provide more growth surface area versus traditional cell culture vessels of comparable footprint. With closed system accessories and multiple size offerings, Corning HYPERStack® cell culture vessels are an efficient, scalable cell culture vessel platform for adherent cell culture and can be used as seed train for adherent cell culture bioreactors.
Application
Adherent cell culture, cell expansion, stem cell expansion, cell banking, cell production, viral vector production, vaccine production
Features and Benefits
  • New updated design
  • Features 6,000 cm2 cell growth area
  • Corning High Yield PERformance (HYPER) gas-permeable film technology
  • Closed system approach with chemically resistant, heat sealable, flexible tubing and female MPC for liquid handling manipulations
  • Corning CellBIND® surface treatment for enhanced cell attachment and expansion
  • Manfactured from USP Class VI virgin polystyrene which provides excellent optical clarity and mechanical strength
  • Sterilized to a sterility assurance level (SAL) 10-6
  • 100% leak tested throughout the manufacturing process
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.