Key Documents
CLS1395OR
Corning® Silicone O-Ring
for use with GL45 Pyrex® Bottles
About This Item
UNSPSC Code:
41121815
sterility
non-sterile
packaging
pack of 10
General description
These silicone O-rings are used in place of the pouring rings on PYREX GL 45 bottles to ensure a leak-resistant seal.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)