Key Documents
CLS10043
Corning® HYPERStack® disposable tubing set
with 850 cm2 polystyrene roller bottle, sterile
Synonym(s):
Corning Tubing Set, Disposable Tubing Set, HYPERStack Tubing Set
About This Item
UNSPSC Code:
41105108
NACRES:
NA.75
description
18 in. C-Flex® tubing
sterility
sterile
packaging
pkg of 2 ea
manufacturer/tradename
Corning 10043
Related Categories
General description
Corning® HYPERStack disposable tubing set is sterile C-Flex® tubing compatible for use with 850 cm2 polystyrene roller bottle. The tubing features PureFit tube clamp system, 0.2 µm filter and MPC connector with cap.
Features and Benefits
  • It has flexible tubing with a heat seal and chemical resistant
  • Easy Grip, threaded cap with male medical grade polycarbonate connector
  • Autoclavable with PureFit®tube clamp system
  • Material is free from animal origin with a polytetrafluoroethylene (PTFE) polymer filter vent
  • Sterilized, with a Sterility Assurance Level (SAL) of 10-6
Legal Information
C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.