Skip to Content

CLL1097

A549 CELLS EGFR BIOSENSOR

human male lung (Source Disease: Lung carcinoma)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Biological source:
human male lung (Source Disease: Lung Carcinoma)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

A549 CELLS EGFR BIOSENSOR,

biological source

human male lung (Source Disease: Lung Carcinoma)

Quality Segment

OMIM accession no.

storage temp.

−196°C

Gene Information

General description

This cell line was derived from ATCC catalog No. CCL-185.

Application

The EGFR Biosensor cell line consists of the SH2 domain of the adaptor protein Grb2 fused to GFP. The biosensor was transduced into the A549 cell line and a stable cell line was cloned from a single cell. Within minutes after stimulation of the cell line with EGF, the biosensor shows robust redistribution towards the cell membrane and subsequent internalization through endocytosis. The internalization kinetics of the sensor-receptor complex can be quantified by image analysis software to count granules. The reporter response is highly selective as it can be induced only by specific EGFR ligands and is abolished by a selective inhibitor of EGFR, Tyrphostin AG 1478.
To learn more, please visit the EGFR Biosensor Cell Line webpage

Features and Benefits

Lentiviral transduction of DNA construct for stable expression of GFP-tagged SH2 binding domain.
These A549 cells are adherent with a doubling time of approx. 22 hours.

Preparation Note

Cell freezing medium-DMSO 1X, Cat. No. C6164
Media renewal changes two to three times per week.
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C.
The base medium for this cell line is RPMI-1640, Cat. No. R0883. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F2442, to a final concentration (v/v) of 10%, L-glutamine, Cat. No. G7513, at a final concentration of 2 mM and puromycin, Cat. No. P9620, at a final concentration of 1 μg/mL.

Analysis Note

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

6.2 - Infectious substances

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library