Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3A(tPA-30-1)

96020926, human placenta, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

3A(tPA-30-1), 96020926

biological source

human placenta

growth mode

Adherent

karyotype

Hypodiploid, modal chromosome number of 42 occuring in 34% of cells. Most cells contained 41-45 chromosomes. Cells with 44 chromosomes also occurred at a high frequency (26%). Cells having a higher ploidly occurred at 20.7%, which is very high.

morphology

Epithelial

products

At 40C cells synthesize human chorionic gonadotrophin (hCG) and alkaline phosphatase.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human placenta, SV40 transformed

Cell Line Description

Established by SV40 ts30 transformation of human placental cells. The cells express the transformed phenotype at the permissive temperature (33C) and the non-transformed phenotype at the non-permissive temperature (40C). The line has a limited life expectancy of 15 to 18 passages before entering the crisis stage.
GMO1

Culture Medium

α-MEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-6x10,000 cells/cm2 using 0.25% trypsin or trypsin or trypsin/EDTA; 5% CO2; 36°C. Change medium 2-3 times weekly.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service