Skip to Content
MilliporeSigma

RPMI 8226

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87012702, human blood, Not specified

Synonym(s):

RPMI8226 Cells, Roswell Park Memorial Institute 8226 Cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human blood
Relevant disease(s):
cancer
Growth mode:
Suspension
Karyotype:
Not specified
Morphology:
Not specified
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human blood

packaging

tube of 5 μg 87012702-DNA-5UG, pkg of vial of cells 87012702-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Immunoglobulin G (Ig) lambda light chain

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

RPMI 8226 has been used:
  • to grow human myeloma cell lines
  • to evaluate the effect of 1-aryl-2-methylthioimidazolines (IIV) on the viability of RPMI 8226 cells
  • to measure cytokine production by bone marrow stromal cells (BMSCs) after contact with the RPMI 8226 cell line

Biochem/physiol Actions

Derived from the peripheral blood of a 61 year old male with multiple myeloma. The cells produce and secrete Ig lambda light chain.
Human myeloma
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 12
D13S317: 11
D16S539: 9
D5S818: 11,13
D7S820: 9,10
THO1: 8
TPOX: 8,11
vWA: 16,18

Preparation Note

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. After resuscitation cell debris will be present in culture. This will decrease with subsequent subculture.
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library