Skip to Content
MilliporeSigma
All Photos(1)

Documents

3T3 clone A31 Cell Line from mouse

86110401, mouse embryo, Fibroblast

Synonym(s):

3T3-A31 Cells, BALB/3T3 clone A31 Cells, BALB/c 3T3 clone A31 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

3T3 clone A31 Cell Line from mouse, 86110401

biological source

mouse embryo

description

Mouse BALB/c embryo

growth mode

Adherent

karyotype

2n = 40, hypotetraploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c embryo

Cell Line Description

Cells have now lost their contact inhibition.

Application

3T3 clone A31 cell line from mouse has been used:
  • to study in vitro cytotoxicity and genotoxicity of retinyl palmitate–loaded poly(lactide-co-glycolide) nanoparticles for the topical treatment of skin diseases
  • for the characterization of the phototoxicity of Vemurafenib, which is a B-Raf serine/threonine kinase inhibitor
  • for the phototoxicity assessment of systemically applied compounds and drug candidates
Tumourigenicity and virus studies: SV40 and MSV

Culture Medium

DMEM + 2mM Glutamine + 5% New Born Calf Serum (NBCS) + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:20 i.e. seeding at 5x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service