Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MCC13 cell line human

10092302, human skin, Large, low contrast cells

Synonym(s):

Human MCC13 Line, MCC13 Human Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MCC13 cell line human, 10092302

biological source

human skin

description

Merkel carcinoma from human skin

growth mode

Not specified

karyotype

Not specified

morphology

Large, low contrast cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Merkel carcinoma

Cell Line Description

Merkel cell carcinoma cell line. Cell Bank Australia reference number: CBA1338

Culture Medium

RPMI 1640 (with 2mM L-Glutamine+25mM HEPES) + 15% FCS

Subculture Routine

Optimal split ratio 1:8 (seeding density 0.8 x104 cells/cm2).Cells incubated at 37°C with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Helen Leonard - Queensland Institute of Medical Research, Australia

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service