Skip to Content
MilliporeSigma

C9749

CBiPES hydrochloride

≥98% (HPLC)

Synonym(s):

N-(4′-cyano[1,1′-biphenyl]-3-yl)-N-(3-pyridinylmethyl)-ethanesulfonamide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H19N3O2S · HCl
CAS Number:
Molecular Weight:
413.92
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to light brown

solubility

DMSO: >20 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N(Cc3cnccc3)c1cc(ccc1)c2ccc(cc2)C#N)CC.Cl

InChI

1S/C21H19N3O2S.ClH/c1-2-27(25,26)24(16-18-5-4-12-23-15-18)21-7-3-6-20(13-21)19-10-8-17(14-22)9-11-19;/h3-13,15H,2,16H2,1H3;1H

InChI key

QKUHSVQWWZBAKM-UHFFFAOYSA-N

Biochem/physiol Actions

CBiPES is a positive allosteric modulator of the mGlu2 receptor.
CBiPES is a positive allosteric modulator of the mGlu2 receptor. CBiPES attenuates stress-induced hyperthermia and blocks the hyperlocomotor effects of PCP in murine models. In rat brain, CBiPES attenuates ketamine-induced increase in extracellular histamine concentration. This compound is a sulfonamide, different chemical class from other mGlu2R agonists in the Sigma catalog.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library