Skip to Content
MilliporeSigma

C8822

Crocein Scarlet 7B

Powder

Synonym(s):

Acid Red 71

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C24H18N4O7S2Na2
CAS Number:
Molecular Weight:
584.53
Colour Index Number:
27165
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Crocein Scarlet 7B,

form

powder

Quality Segment

composition

Dye content, ~65-85%

color

red to brown

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na].Cc1cc(ccc1\N=N\c2c(O)ccc3cccc(c23)S(O)(=O)=O)\N=N\c4ccc(cc4C)S(O)(=O)=O

InChI

1S/C24H20N4O7S2.Na.H/c1-14-12-17(25-26-20-10-8-18(13-15(20)2)36(30,31)32)7-9-19(14)27-28-24-21(29)11-6-16-4-3-5-22(23(16)24)37(33,34)35;;/h3-13,29H,1-2H3,(H,30,31,32)(H,33,34,35);;/b26-25+,28-27+;;

InChI key

HJEUGBJDHUTXMF-NPYGFOBASA-N

Biochem/physiol Actions

Crocein Scarlet 7B (acid red 71) is used in Crowle′s reagent. The reagent is needed for observing latent fingerprints, particularly bloody prints.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library