Skip to Content
MilliporeSigma

C8051

Collagenase from Clostridium histolyticum

Sigma Blend Type H, ≥1.0 FALGPA units/mg solid

Synonym(s):

Clostridiopeptidase A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-582-9
MDL number:
EC Number:
Specific activity:
≥1.0 FALGPA units/mg solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

caseinase activity

≥10 units/mg solid

specific activity

≥1.0 FALGPA units/mg solid

mol wt

68-130 kDa

storage temp.

−20°C

Application

Collagenase from Clostridium histolyticum has been used:
  • to harvest osteosarcoma 143B cells from collagen scaffold
  • in tissue digestion of islets and carotid artery
  • in the digestion of chorionic villi
  • in enzymatic dissociation for ATP-based tumour chemosensitivity assay (ATP-TCA) in solid tumors

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of both collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. The pH optimum is 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Collagenase treatment can cause some cells to die. Typically, concentrations varying from 0.1 to 5 mg/mL is used for digestion. The duration of reaction varyies from 15 minutes to several hours to yield a satisfactory efficiency of cell dissociation, without causing too much cell death. Krebs Ringer buffer with calcium and BSA is preffered and Zn2+ is required for activity. This enzyme is tested for the release of hepatocytes at approximately 1 mg/mL conc., in a total volume of 100 mL for each rat liver.
Clostridium histolyticum produces two classes of collagenases. Clostridial collagenases are found to be highly effective than mammalian collagenases. It is capable of cleaving at multiple cleavage sites within the collagen triple helix. It is implicated in the bacterial invasion in gas gangrene.

Other Notes

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library