Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6249

Sigma-Aldrich

Catestatin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

lyophilized powder

Quality Level

solubility

deionized water: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CHGA(1113)

Amino Acid Sequence

Ser-Ser-Met-Lys-Leu-Ser-Phe-Arg-Ala-Arg-Ala-Tyr-Gly-Phe-Arg-Gly-Pro-Gly-Pro-Gln-Leu

Biochem/physiol Actions

The catestatin peptide is produced from its precursor chromogranin A protein. Catestatin is a cationic and hydrophobic peptide that can stimulate histamine release from mast cells and exhibits potent catecholamine relase-inhibitory activity.

Physical form

Lyophilized from 0.1% aqueous TFA.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service