Skip to Content
MilliporeSigma

C6187

Nalgene® sealing cap assembly

diam. 24 mm, pack of 2 ea

Synonym(s):

bottle seal, tube sealing device, Nalgene DS3131-0024, plastic cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.11
UNSPSC Code:
41121703
Material:
Tefzel® plug, Viton® O-ring, polypropylene screw closure
Packaging:
pack of 2 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

Tefzel® plug, Viton® O-ring, polypropylene screw closure

packaging

pack of 2 ea

manufacturer/tradename

Nalgene DS3131-0024

diam.

24 mm

Application

Use with recommended tubes and bottles for leakproof service at maximum speed. Also recommended for spinning hazardous materials.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company


Still not finding the right product?

Explore all of our products under Nalgene® sealing cap assembly




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library