Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5613

Sigma-Aldrich

Anti-Calcium Channel (α2/δSubunit) antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL using rat brain tissue lysate

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The calcium channel α2/δ subunit is a glycosylated structural subunit consisting of the α2 subunit and the δ peptide.

Specificity

Specifically recognizes a splice variant of α2/δ (122 kDa).

Immunogen

synthetic peptide derived from the rat α2/δ calcium channel subunit conjugated to KLH.

Application

Anti-Calcium Channel α2/δ subunit is suitable for immunoblotting and western blotting at a concentration of 5-10μg/mL using rat brain tissue lysate.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service