Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5467

SAFC

EX-CELL® ACF CHO Medium

Animal-component free, with HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

CHO Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

quality

Drug/Device Master File available

technique(s)

cell culture | mammalian: suitable

components

HEPES: yes
NaHCO3: yes
phenol red: no
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Animal component-free medium formulated to optimize cell growth and protein expression in Chinese hamster ovary (CHO) cells.

Features and Benefits

Developed to meet the needs of biotechnology and vaccine manufacturing, this medium supports rapid initial cell growth and high levels of protein expression in suspension cultures. It also supports high cell densities and maintenance of these densities of viable cells for extended periods resulting in increased productivity. Cell densities in excess of 8 × 106 cells/ml have been achieved in batch culture systems. Increases in protein productivity up to 500% per cell have been observed.

Other Notes

Proprietary formulation containing inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, recombinant human insulin, plant hydrolysates, other organic compounds, trace elements, and surfactants.
Does not contain antibiotics, antimycotics, L-glutamine, or transferrin. Contains no animal-derived proteins or other components.

Reconstitution

Aseptically add 20-40 ml of 200 mM L-glutamine solution per liter of medium prior to use.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Silvia Gaggero et al.
Oncoimmunology, 7(9), e1470730-e1470730 (2018-09-20)
The release of soluble ligands of activating Natural Killer (NK) cell receptors may represent a regulatory mechanism of NK cell function both in physiologic and in pathologic conditions. Here, we identified the extracellular matrix protein Nidogen-1 (NID1) as a ligand
Jonas Goldin Diness et al.
Frontiers in pharmacology, 11, 749-749 (2020-06-09)
Hypokalemia reduces the cardiac repolarization reserve. This prolongs the QT-interval and increases the risk of ventricular arrhythmia; a risk that is exacerbated by administration of classical class 3 anti-arrhythmic agents.Small conductance Ca2+-activated K+-channels (KCa2) are a promising new atrial selective
Chris Chambers et al.
Assay and drug development technologies, 14(2), 93-108 (2016-03-19)
Voltage-gated sodium (Na(V)) channels have an essential role in the initiation and propagation of action potentials in excitable cells, such as neurons. Of these channels, Na(V)1.7 has been indicated as a key channel for pain sensation. While extensive efforts have

Articles

Cell culture protocol for passaging and splitting suspension cell lines

MGAT1 adds N-acetylglucosamine to the Man5GlcNAc2 (Man5) structure. Goh et al. reported increased sialylation after restoring MGAT1 function in MGAT1 deficient CHO cells.

A signal peptide is a 5-30 amino acid (aa) peptide present at the N-terminus of secretory proteins.

In the present study, we have identifi ed species-specifi c housekeeping genes (HKGs) for Chinese Hamster Ovary (CHO) cells using data from microarray gene expression profiling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service