Skip to Content
MilliporeSigma

C5236

CelLytic IB Inclusion Body Solubilization Reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
41116134
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

storage temp.

room temp

Application

CelLytic IB was designed to solubilize protein aggregates called inclusion bodies, which are formed in bacteria when recombinant proteins are overexpressed. CelLytic IB was formulated to solubilize the protein of interest for immediate analysis of protein content or refolding procedures.

Legal Information

CelLytic is a trademark of Sigma-Aldrich Co. LLC


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library