Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C5041

Sigma-Aldrich

6-Carboxyfluorescein diacetate

~95% purity (HPLC), powder

Synonym(s):

6-CFDA, 6-Carboxy-di-O-acetylfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16O9
CAS Number:
Molecular Weight:
460.39
Beilstein:
67043
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

6-Carboxyfluorescein diacetate, ≥95% (HPLC)

Quality Level

100
200

Assay

≥95% (HPLC)

form

powder

color

white to yellow

solubility

chloroform/ethanol (1:1): 9.80-10.20 mg/mL, clear, colorless to light yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5ccc(cc45)C(O)=O)c1

InChI

1S/C25H16O9/c1-12(26)31-15-4-7-18-21(10-15)33-22-11-16(32-13(2)27)5-8-19(22)25(18)20-9-14(23(28)29)3-6-17(20)24(30)34-25/h3-11H,1-2H3,(H,28,29)

InChI key

QMOGCCYGOPYYNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Carboxyfluorescein diacetate has been used to differentiate viable cells from apoptotic cells. Live cells that are not apoptotic accumulate 6-carboxyfluorescein but do not accumulate annexin.
Fluorescent wavelengths λex 492nm; λemm 517nm in 0.1M Tris, pH 8.0.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of sperm quality snakes Erythrolamprus poecilogyrus sublineatus (Cope, 1860) (Serpentes, Dipsadidae).
Silva AC, et al.
Brazilian journal of biology = Revista brasleira de biologia, doi: 10-doi: 10 (2017)
Gopal Patel et al.
Frontiers in bioengineering and biotechnology, 8, 631-631 (2020-07-03)
The present study explores the influence of mycophenolic acid (MPA) in combination therapy with quercetin (QC) (impeding MPA metabolic rate) delivered using the liposomal nanoparticles (LNPs). Mycophenolic acid liposome nanoparticles (MPA-LNPs) and quercetin liposome nanoparticles (QC-LNPs) were individually prepared and
S Capellero et al.
Scientific reports, 10(1), 6478-6478 (2020-04-17)
Despite improvements in surgery and medical treatments, epithelial ovarian cancer (EOC) remains the most lethal gynaecological malignancy. Aim of this study is to investigate the preclinical immunotherapy activity of cytokine-induced killer lymphocytes (CIK) against epithelial ovarian cancers, focusing on platinum-resistant
G Ricci et al.
Human reproduction (Oxford, England), 17(10), 2665-2672 (2002-09-28)
Apoptosis plays an important role in regulating spermatogenesis. However, the biological significance of apoptosis in ejaculated sperm is not yet clear. This study set out to investigate how apoptosis correlates with semen quality and the presence of seminal leukocytes. Fifty-seven
Garry R Smith et al.
Journal of molecular neuroscience : MN, 52(3), 446-458 (2013-11-28)
Severe seizure activity is associated with reoccurring cycles of excitotoxicity and oxidative stress that result in progressive neuronal damage and death. Intervention with these pathological processes is a compelling disease-modifying strategy for the treatment of seizure disorders. We have optimized

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service