Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C4832

Sigma-Aldrich

Ceramide 1-phosphate from bovine brain

~95%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Assay

~95%

Quality Level

form

powder

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)N[C@@H](COP(O)(O)=O)[C@H](O)\C=C\CCCCCCCCCCCCC

InChI

1S/C36H72NO6P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-36(39)37-34(33-43-44(40,41)42)35(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,34-35,38H,3-28,30,32-33H2,1-2H3,(H,37,39)(H2,40,41,42)/b31-29+/t34-,35+/m0/s1

InChI key

ZQQLMECVOXKFJK-NXCSZAMKSA-N

Related Categories

Biochem/physiol Actions

Putative second messenger of agonist-stimulated sphingomyelin metabolism; may participate in cell regulation and transmembrane signaling.

Other Notes

Contains primarily stearic and nervonic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Shinghal et al.
Journal of neurochemistry, 61(6), 2279-2285 (1993-12-01)
Recent studies have implicated sphingolipids in a variety of intracellular signaling systems. The finding that a calcium-stimulated ceramide kinase copurifies with neurotransmitter-containing vesicles suggests that ceramide, or one of its metabolites, has a role in neurotransmitter release. As a step
O Boudker et al.
The Journal of biological chemistry, 268(29), 22150-22155 (1993-10-15)
A calcium-dependent ceramide (Cer) kinase was recently detected in human leukemia (HL-60) cells (Kolesnick, R.N., and Hemer, M.R. (1990) J. Biol. Chem. 265, 18803-18808) where it may function in terminating the regulatory effects of Cer, and in synaptic vesicles (Bajjalieh
Shaowei Wang et al.
Molecular neurodegeneration, 16(1), 26-26 (2021-04-18)
Apolipoprotein E4 (APOE4) is associated with a greater response to neuroinflammation and the risk of developing late-onset Alzheimer's disease (AD), but the mechanisms for this association are not clear. The activation of calcium-dependent cytosolic phospholipase A2 (cPLA2) is involved in
Chinmoy Sarkar et al.
Autophagy, 16(3), 466-485 (2019-06-27)
Lysosomal membrane permeabilization (LMP) is observed under many pathological conditions, leading to cellular dysfunction and death. However, the mechanisms by which lysosomal membranes become leaky in vivo are not clear. Our data demonstrate that LMP occurs in neurons following controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service