Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

C4780

Sigma-Aldrich

Coenzyme A sodium salt hydrate

BioReagent, suitable for cell culture

Synonym(s):

CoA Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36N7O16P3S · xNa+ · yH2O
CAS Number:
Molecular Weight:
767.53 (anhydrous free acid basis)
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

yeast

description

cofactor for acyl transfer

product line

BioReagent

Assay

≥85% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na+].CC(C)(COP(O)(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)([O-])=O)n2cnc3c(N)ncnc23)C(O)C(=O)NCCC(=O)NCCS

InChI

1S/C21H36N7O16P3S.Na/c1-21(2,16(31)19(32)24-4-3-12(29)23-5-6-48)8-41-47(38,39)44-46(36,37)40-7-11-15(43-45(33,34)35)14(30)20(42-11)28-10-27-13-17(22)25-9-26-18(13)28;/h9-11,14-16,20,30-31,48H,3-8H2,1-2H3,(H,23,29)(H,24,32)(H,36,37)(H,38,39)(H2,22,25,26)(H

InChI key

SYTRWOCXZXQBPW-CLVRNSBASA-M

Looking for similar products? Visit Product Comparison Guide

Application

Coenzyme A sodium salt hydrate has been used:
  • in the branch chain aminotransferase enzymatic assay and protein determination
  • in the assay for halogenation of pyrrolyl-S-Bmp1 (an acyl carrier protein (ACP)-thioesterase (TE) di-domain protein) by Bmp2
  • as an extracellular coenzyme A (CoA) in in vitro and in vivo experiments to study alternative routes by which cells and organisms can obtain CoA

Biochem/physiol Actions

Coenzyme A (CoA) is a ubiquitous cofactor that plays an important role in many metabolic pathways, mainly as acyl-CoA. It is essential for the tricarboxylic acid cycle, and the synthesis and oxidation of fatty acids. CoA facilitates acyl group transfers, especially in fatty acid and lipid metabolism reactions. It is associated with neurodegeneration, autophagy, protein acetylation, and signal transduction.

Quality

Enzymatically assayed using phosphotransacetylase. Approx. 95% CoA-SH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balaji Srinivasan et al.
Nature chemical biology, 11(10), 784-792 (2015-09-01)
The metabolic cofactor coenzyme A (CoA) gained renewed attention because of its roles in neurodegeneration, protein acetylation, autophagy and signal transduction. The long-standing dogma is that eukaryotic cells obtain CoA exclusively via the uptake of extracellular precursors, especially vitamin B5
Yongzhong Wei et al.
World journal of gastroenterology, 14(2), 193-199 (2008-01-12)
Nonalcoholic fatty liver disease (NAFLD) includes hepatic steatosis, nonalcoholic steatohepatitis (NASH), fibrosis, and cirrhosis. NAFLD is the most common liver disorder in the United States and worldwide. Due to the rapid rise of the metabolic syndrome, the prevalence of NAFLD
Yao Rong et al.
eLife, 10 (2021-08-20)
TMEM120A, a member of the transmembrane protein 120 (TMEM120) family, has a pivotal function in adipocyte differentiation and metabolism, and may also contribute to sensing mechanical pain by functioning as an ion channel named TACAN. Here we report that expression
Le Tran Phuc Khoa et al.
Cell stem cell, 27(3), 441-458 (2020-07-02)
Self-renewing embryonic stem cells (ESCs) respond to environmental cues by exiting pluripotency or entering a quiescent state. The molecular basis underlying this fate choice remains unclear. Here, we show that histone acetyltransferase MOF plays a critical role in this process
Krista Yang et al.
STAR protocols, 2(3), 100687-100687 (2021-08-18)
Carnitine palmitoyltransferase-1 (CPT-1) is a rate-controlling enzyme for long-chain fatty acid oxidation. This manuscript provides protocols for measuring CPT-1-mediated respiration in permeabilized, adherent cell monolayers and mitochondria freshly isolated from tissue, along with examples to assess the potency and specificity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service