Skip to Content
MilliporeSigma

Skip To

C4361

Collagen Type I−FITC Conjugate from bovine skin

~1 mg/mL, solution

Synonym(s):

Collagen from bovine skin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine skin

Quality Segment

form

solution

concentration

~1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

General description

Type I collagen is the major collagen of tendon and bone, but it is widely distributed in other tissues, such as skin, cornea, tooth dentin, heart valve, lung, liver, fascia and scar tissue.

Application

Collagen Type I-FITC Conjugate from bovine skin has been used in collagen I gel deformation analysis. It has also been used to coat silicone elastic membranes for seeding cells for various experimental studies. It can also be used as a substrate for assaying mammalian collagenase activity in cell culture.

Biochem/physiol Actions

Collagen type I offers mechanical stability, strength and toughness to a range of tissues from tendons and ligaments, to skin, cornea, bone and dentin.

Physical form

Supplied in 0.01 M acetic acid.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C4243234138C9879
description

~1 mg/mL, solution

description

2.9-3.2 mg/mL, (Suitable for cell culture and for 3D matrix formation), sterile-filtered

description

-

description

powder, suitable for substrate for collagenase

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

solution

form

liquid

form

lyophilized

form

powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

ambient

shipped in

-

Gene Information

cow ... COL1A1(282187), COL1A2(282188)
human ... MMP1 (includes EG:4312)(4312)
mouse ... MMP1 (includes EG:4312)(83995)
rat ... MMP1 (includes EG:4312)(300339)

Gene Information

human ... COL1A1(282187)

Gene Information

-

Gene Information

-

biological source

bovine skin

biological source

bovine skin

biological source

human skin

biological source

bovine Achilles tendon


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Faceshields, Gloves, Goggles



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters