Key Documents
C3983
Sigma-Aldrich
Scienceware® cloning cylinders, polystyrene
I.D. × H 6.4 mm × 8 mm
Synonym(s):
cloning cylinder, cloning equipment
About This Item
UNSPSC Code:
41106618
NACRES:
NB.22
sterility
sterile
packaging
pack of 50 ea
manufacturer/tradename
Bel-Art F37847-0200
I.D. × H
6.4 mm × 8 mm
General description
To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)