Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-39216

Sigma-Aldrich

Endothelial Cell Growth Medium 2

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-22211

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Endothelial Cell Growth Medium 2 SupplementMix (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-22211

Application

Endothelial Cell Growth Medium is a low-serum (2% V/V) medium optimized for the cultivation of endothelial cells from large blood vessels. The media are optimized for primary human cells but can also be used for bovine, porcine, murine, caprine, rabbit, elephant, dog, and rat endothelial cells from large vessels as well as for various endothelial cell lines.Note: For best transfection results we recommend using PromoCell heparin-free Endothelial Cell Growth Medium MV2. Heparin, included all our other our Endothelial Cell Growth Media formulations, may reduce transfection efficiency.

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Endothelial Cell Media 2 are subjected to comprehensive quality control tests using primary human endothelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested endothelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Epidermal Growth Factor (recombinant human): 5 ng / mlBasic Fibroblast Growth Factor (recombinant human): 10 ng / ml Insulin-like Growth Factor (Long R3 IGF): 20 ng / ml Vascular Endothelial Growth Factor 165 (recombinant human): 0.5 ng / mlAscorbic Acid: 1 µg / mlHeparin: 22.5 µg / ml Hydrocortisone: 0.2 µg / ml

Storage and Stability

Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.

Related Content

Cell culture protocol: the endothelial tube formation assay to study angiogenesis using HUVECs and other endothelial cell types. Explore over 350 PromoCell products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service