Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352202
Quality Segment
form
liquid
usage
sufficient for 100-200 applications (on a PhastGel (Cytiva))
mol wt
6,500-45,000 Da
shipped in
dry ice
storage temp.
−70°C
Packaging
Each vial contains 0.5 mL of a solution of five colored dye conjugated proteins.
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.