Skip to Content
MilliporeSigma

C3124

CryoStor® cell cryopreservation media

CS2

Synonym(s):

Cell Cryopreservation Solution, Cryopreservation Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable, cryopreservation: suitable

shipped in

ambient

storage temp.

2-8°C

General description

The CryoStor CS2, CS5, and CS10 family of preservation solutions represents the next generation of cryopreservation media. Designed to prepare and preserve cells in ultra low temperature environments (-80 to -196 °C), CryoStor media provide a safe, protective environment for cells and tissues during thefreezing, storage, and thawing process. Through modulating the cellular biochemical response to the cryopreservation process, these media provide enhanced cell viability and functionality, while eliminating the need to include serum, proteins, or high levels of cytotoxic agents.

CryoStor CS2, CS5, and CS10 are a series of cell specific, optimized preservation media, uniquely formulated to address the molecular biological aspects of cells during the cryopreservation process; thereby, directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.

These media are recommended for the preservation of stem cells, hepatocytes, tissue samples, and other extremely sensitive cell types.

Application

CryoStor CS2 is formulated to contain 2% dimethyl sulfoxide (DMSO). Suggested when reducing DMSO is of primary concern.

Cryostor® Cryopreservation Video Protocol
CryoStor, a series of cell-specific, optimized preservation media, is uniquely formulated to address the molecularbiological aspects of cells during the cryopreservation process thereby directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.
CryoStor® cell cryopreservation media has been used in the cryopreservation of Plasmodium vivax and P. falciparum sporozoites.

Other Notes

Formulation contains 2% DMSO.

Legal Information

CryoStor is a registered trademark of BioLife Solutions, Inc.


Still not finding the right product?


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library