Skip to Content
MilliporeSigma

Skip To

C-28077

Cancer Cell Line Medium XF

250 ml

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.81
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

PromoCell®

form

liquid

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Cancer Cell Line Medium XF

Application

Cancer Cell Line Medium XF is a cell culture medium developed for the standardized in vitro cultivation of established human cancer cell lines. Its serum-free and xeno-free formulation provides a culture environment devoid of all stimuli originating from non-defined materials. Thus, it has no ill-defined components such as fetal calf serum, extracts or hydrolysates and exhibits very low lot-to-lot variability. The medium consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete medium.

Preparation Note

Click here for more information.
Store the Basal Medium at 2 to 8°C in the dark, store the SupplementMix at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 2 to 8°C. For use, prewarm only an aliquot of the complete medium and keep the remaining medium refrigerated at 2 to 8°C.

Analysis Note

All lots of the PromoCell Cancer Cell Line Medium XF are subjected to comprehensive quality control tests using a human cancer cell line. Each lot of PromoCell Cancer Cell Line Medium XF is tested for the ability to support proliferation and serial passage of MCF7 breast carcinoma cells on fibronectincoated dishes. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Legal Information

PromoCell is a registered trademark of PromoCell GmbH


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Would I be able to find out the glucose, cholesterol and lipid concentrations in this media? I am doing an experiment where these need to be carefully titrated, and I need to know the starting amounts. Thank you.

    1 answer
    1. The formulation of this medium is proprietary, and therefore, information about glucose, cholesterol, and lipid concentrations cannot be provided.

      Helpful?

Reviews

No rating value

Active Filters