Skip to Content
MilliporeSigma

C-28053

Dendritic Cell Generation Medium

Basal Medium, 250 ml

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

DC Base Medium (supplements sold separately)

Application

The Dendritic Cell (DC) Base Medium is a user-customizable version of the DC Generation Media that does not include cytokines. It must be adequately supplemented by the user for the generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes.Note: DC Base Medium must be supplemented with additional cytokines. For freshly isolated cells, the Monocyte Attachment Medium (C-28051) is also needed.Recommended for:Human Monocytes (hMo), freshly isolated and cryopreservedHuman Mononuclear Cells (hMNC), freshly isolated

Preparation Note

Click here for more information.
Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the SupplementMix to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Complete Medium supplemented with cytokines should be used within 2 weeks. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Analysis Note

All lots of PromoCell DC Media are subjected to comprehensive quality control tests using human peripheral blood mononuclear cells. Each lot of PromoCell DC Media is tested for the ability to support DC maturation verified by morphological evaluation and flow cytometry analysis. Each lot of PromoCell Monocyte Attachment Medium is tested for its ability to support optimal attachment and viability of peripheral blood Monocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).


Still not finding the right product?

Explore all of our products under Dendritic Cell Generation Medium


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library