Skip to Content
MilliporeSigma

C2360

CelLytic P Cell Lysis Reagent

Synonym(s):

Plant cell lysis reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
41116134
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For plant cell lysis

Quality Segment

form

solution

storage temp.

room temp

Application

CelLytic P Cell Lysis Reagent has been used for lysate preparation for cell lysis.

Other Notes

CelLytic P is a non-ionic detergent-based reagent, which offers a convenient method for efficient plant cell lysis and protein solubilization. It is a non-denaturing reagent and maintains protein immunoreactivity and biological activity. CelLytic P is efficient, rapid, and ready to use. It contains bicine buffer, which is preferable for many biological activities.

Legal Information

CelLytic is a trademark of Sigma-Aldrich Co. LLC


pictograms

Exclamation mark

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library