Skip to Content
MilliporeSigma

C1934

Magenta 2-way cap

natural

Synonym(s):

test tube cap, test tube caps

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41121800
Color:
natural
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


color

natural

compatibility

for use with 25 mm culture tubes

General description

  • Magenta 2-way cap is made of polypropylene.
  • 25 mm diameter x 31 mm height.
  • Works in conjunction with 25 mm Test Tubes.
  • Drip Ring minimizes contamination by preventing condensation between the test tube and cap wall.
  • Cap groove accommodates filter to eliminate contamination by purifying incoming air.
  • Flexible ribs.
  • Visually transparent.
  • Autoclavable and reusable.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Global Trade Item Number

SKUGTIN
C1934-100EA04061833302033
C1934-500EA04061833574867