Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Recombinant:
expressed in E. coli
Concentration:
≥12 U/mL
recombinant
expressed in E. coli
Quality Level
form
ammonium sulfate suspension
concentration
≥12 U/mL
storage temp.
−20°C
Application
Cyclohexanone monooxygenase has been used in a study that cloned and overexpressed the 2-oxo-Δ(3)-4,5,5-trimethylcyclopentenylacetyl-CoA monooxygenase (OTEMO) in Escherichia coli. Cyclohexanone monooxygenase has also been used in a study that investigated the effects of structural modification of the cell wall on the biotransformation capability by recombinant Corynebacterium glutamicum cells.
Biochem/physiol Actions
Purified cyclohexanone monooxygenase has a molecular weight of 59,000 Da. It is a versatile oxygenation catalyst. The enzyme uses the bound FAD-4a-OOH oxygenating intermediate to initiate transfer of oxygen to electrophilic substrate sites. The reaction consequently yields the corresponding sulfoxide and selenoxide products. This enzyme is also capable of oxygenating at nitrogen, trivalent phosphorus, and boron sites in boronic acids. Hence, it is one of the most broad-based flavoprotein oxygenases known.
Physical form
Suspension in 80% saturated ammonium sulfate, 20 mM K-Na-phosphate buffer pH 7, 3.5 mM 1,4-Dithioerythritol (DTE)
Other Notes
1 unit corresponds to the amount of enzyme which catalyzes the cyclohexanone-stimulated oxidation of 1 μmol of NADPH per minute at pH 9.0 and 30 °C
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Y C Chen et al.
Journal of bacteriology, 170(2), 781-789 (1988-02-01)
The gene coding for cyclohexanone monooxygenase from Acinetobacter sp. strain NCIB 9871 was isolated by immunological screening methods. We located and determined the nucleotide sequence of the gene. The structural gene is 1,626 nucleotides long and codes for a polypeptide
Hannes Leisch et al.
Applied and environmental microbiology, 78(7), 2200-2212 (2012-01-24)
A dimeric Baeyer-Villiger monooxygenase (BVMO) catalyzing the lactonization of 2-oxo-Δ(3)-4,5,5-trimethylcyclopentenylacetyl-coenzyme A (CoA), a key intermediate in the metabolism of camphor by Pseudomonas putida ATCC 17453, had been initially characterized in 1983 by Ougham and coworkers (H. J. Ougham, D. G.
D Sheng et al.
Biochemistry, 40(37), 11156-11167 (2001-09-12)
Cyclohexanone monooxygenase (CHMO), a bacterial flavoenzyme, carries out an oxygen insertion reaction on cyclohexanone to form a seven-membered cyclic product, epsilon-caprolactone. The reaction catalyzed involves the four-electron reduction of O2 at the expense of a two-electron oxidation of NADPH and
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C1622-1ML | 04061832729015 |
