Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human skeletal muscle
Morphology:
(skeletal muscle)
biological source
human skeletal muscle
packaging
pkg of 1,000,000 cells
morphology
(skeletal muscle)
application(s)
cell analysis (DNA, RNA, and protein)
shipped in
dry ice
storage temp.
−20°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.
Application
Primary Human Skeletal Muscle Cells (SkMC) are isolated from different skeletal muscles (e.g. Musculus pectoralis major) from adult single donors (lot specific source information is available on request). They are positive for sarcomeric myosin and negative for smooth muscle specific α-actin.New skeletal muscle cells originate from quiescent satellite cells, which are located in the muscle fibers between the basal lamina and the sarcolemma. Quiescent satellite cells are activated by stimuli such as muscle damage. After activation, the cells, now called myoblasts, start to proliferate and fuse with damaged muscle fibers or with one another forming new myotubes.SkMC are optimal for in vitro muscle studies. They proliferate very well in the mitogen-rich PromoCell Skeletal Muscle Cell Growth Medium. Fusion to myotubes with typical multinucleated syncytia can be induced by using the PromoCell Skeletal Muscle Cell Differentiation Medium.
Biochem/physiol Actions
Muscle
Physical form
Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.
Legal Information
RNAlater is a registered trademark of Thermo Fisher Scientific, Inc.
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
Still not finding the right product?
Explore all of our products under Human Skeletal Muscle Cells (SkMC)
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.